Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium ICP Standard, 1mL = 10mg Cd (10,000ppm Cd), Ricca Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly natural
7440-43-9,7697-37-2,7440-43-9
Cd
112.414
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
CHEBI:22977
cadmium
[Cd]
7440-43-9,7697-37-2,7440-43-9
Cd
112.414
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
CHEBI:22977
cadmium
[Cd]
CAS Min % 87.0,9.84,0.87
CAS Max % 91.0,10.24,0.91
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 10,000ppm Cd
Equivalence 1 mL = 10 mg
Color Colorless
Quantity 500mL
Chemical Name or Material Cadmium ICP Standard