Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Iodide, Crystal, 99%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
7790-80-9
7790-80-9
CAS Max % 1
Percent Purity 0.99
Grade Reagent
Chloride and Bromide 5e-05
Insoluble Matter 5e-05
Iron (Fe) 1e-05
Lead (Pb) 5e-05
Packaging Amber Glass Bottle
Sulfate 5e-05
Zinc (Zn) 0.0005
Quantity 500 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.