Alfa Aesar™ Cadmium iodide, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4344222

Quantity 100g

 View more versions of this product

Catalog No. AA4344222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-80-9
    Molecular Formula CdI2
    Formula Weight 366.21
    MDL Number MFCD00010920
    UN Number UN2570
    Merck Index 14,1621
    Chemical Name or Material Cadmium iodide
    Grade Puratronic™

    Specifications

    Formula Weight 366.21
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P301+P310-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    F SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00010920
    CAS 7790-80-9
    UN Number UN2570
    Merck Index 14,1621
    Health Hazard 2 GHS H Statement
    H301-H331-H351-H373
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    Molecular Formula CdI2
    Boiling Point 787°C
    Color Pale brown
    Quantity 100g
    RTECS Number EV1290000
    TSCA TSCA
    Sensitivity Light Sensitive & Hygroscopic
    Melting Point 387° to 389°C
    Health Hazard 1 Danger
    Density 5.67g/mL
    Chemical Name or Material Cadmium iodide
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H301-H331-H351-H373
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P281-P301+P310-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    F SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit