Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cadmium iodide, ultra dry, 99.9985% (metals basis)

Manufacturer:  Alfa Aesar™ 3568306

Quantity 5g

 View more versions of this product

Catalog No. AA3568306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7790-80-9
MFCD00010920
14,1621
366.21
UN2570
Cadmium iodide, Ultra dry

Specifications

7790-80-9
366.21
UN2570
5.67g/mL
Odorless
Light Sensitive & Hygroscopic
White to yellow
5g
99.9985%, (metals basis)
Beads
MFCD00010920
14,1621
−10 Mesh
Ampule
787°C
387° to 389°C
Cadmium iodide, Ultra dry

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P281-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

RTECSNumber : EV1290000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit