Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cadmium Metal
SCI_ED

Click to view available options
Quantity:
100g
Packaging:
Solid
This item is not returnable. View return policy
7440-43-9
Cd
112.41
MFCD00010914
WLZRMCYVCSSEQC-UHFFFAOYSA-N
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
23973
CHEBI:22977
cadmium(2+) ion
[Cd++]
This item is not returnable. View return policy
7440-43-9
Cd
112.41
MFCD00010914
WLZRMCYVCSSEQC-UHFFFAOYSA-N
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
23973
CHEBI:22977
cadmium(2+) ion
[Cd++]

This product is an Educational Grade Chemical — no C of A is available.

Density 8.64g/mL at 25°C
Assay Percent Range >99%
Linear Formula Cd
Quantity 100g
Formula Weight 112.4
Packaging Solid
Chemical Name or Material Cadmium Metal

ShelfLife : 5 years