Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium Metal, Mossy, 99.9%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
CAS Max % | 1 |
Copper (Cu) | 0.00025 |
Quantity | 500 g |
Iron (Fe) | 1.5e-05 |
Lead (Pb) | 6e-05 |
Percent Purity | 0.999 |
Grade | Reagent |
Packaging | Poly Bottle |
Zinc (Zn) | 0.0002 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.