Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Metal, Powder, 99%, Spectrum™ Chemical
SDP

Catalog Number p-7165262
Click to view available options
Quantity:
125 g
500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7440-43-9
Cd
112.41
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
[Cd]
7440-43-9
Cd
112.41
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
[Cd]

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit