Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium Metal, Powder, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
125 g
500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Specifications
CAS Max % | 1 |
Copper (Cu) | 0.00025 |
Quantity | 500 g |
Iron (Fe) | 2e-05 |
Lead (Pb) | 0.00025 |
Percent Purity | 0.99 |
Grade | Reagent |
Packaging | Poly Bottle |
Zinc (Zn) | 0.00035 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.