Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium Metal, Sticks, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
125 g
500 g
Packaging:
Fiber Tube
Specifications
CAS Max % | 1 |
Copper (Cu) | 5e-05 |
Quantity | 500 g |
Iron (Fe) | 1e-05 |
Lead (Pb) | 0.0002 |
Percent Purity | 0.99 |
Grade | Reagent |
Packaging | Fiber Tube |
Zinc (Zn) | 0.001 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.