Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cadmium molybdenum oxide, 99.5% (metals basis)

Electronic and optical applications

Manufacturer:  Alfa Aesar™ 4020218

Quantity 50g

 View more versions of this product

Catalog No. AA4020218


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13972-68-4
MFCD00015997
Cadmium molybdate
272.35
UN2570
Cadmium molybdenum oxide

Specifications

13972-68-4
272.35
UN2570
Slightly soluble in water; Soluble in acid
≈1250°C
Cadmium molybdenum oxide
Powder
MFCD00015997
Cadmium molybdate
Odorless
50g
99.5%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit