Cadmium nitrate tetrahydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1066322

Quantity 100g

 View more versions of this product

Catalog No. AA1066322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10022-68-1
    Physical Form Crystalline
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula Cd(NO3)2·4H2O
    Formula Weight 308.47
    MDL Number MFCD00149626
    Chemical Name or Material Cadmium nitrate tetrahydrate

    Specifications

    Formula Weight 308.47
    Quantity 100g
    Color White
    EINECS Number 233-710-6
    Physical Form Crystalline
    MDL Number MFCD00149626
    Odor Weak
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula Cd(NO3)2·4H2O
    Appearance White crystalline
    CAS 10022-68-1
    Health Hazard 1 H350-H272-H400-H410-H302-H312-H332
    Chemical Name or Material Cadmium nitrate tetrahydrate

    H350-H272-H400-H410-H302-H312-H332

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit