Cadmium oxide, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1066422

Quantity 100g

 View more versions of this product

Catalog No. AA1066422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1306-19-0
    Physical Form Powder
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula CdO
    MDL Number MFCD00010921
    Chemical Name or Material Cadmium oxide

    Specifications

    Quantity 100g
    EINECS Number 215-146-2
    Physical Form Powder
    Density 8.15
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula CdO
    MDL Number MFCD00010921
    CAS 1306-19-0
    Chemical Name or Material Cadmium oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit