Alfa Aesar™ Cadmium phosphate (ortho), 99%

Manufacturer: Alfa Aesar™ 4020109

Quantity 10g

 View more versions of this product

Catalog No. AA4020109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13847-17-1
    Formula Weight 527.14
    MDL Number MFCD00049414
    UN Number UN2570
    Synonym Cadmium orthophosphate
    Chemical Name or Material Cadmium phosphate (ortho)

    Specifications

    Formula Weight 527.14
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2570
    Synonym Cadmium orthophosphate
    Quantity 10g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Physical Form Powder
    MDL Number MFCD00049414
    Solubility Information Insoluble in water
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    TSCA TSCA
    Melting Point ≈1500°C
    CAS 13847-17-1
    Chemical Name or Material Cadmium phosphate (ortho)
    Assay 99%

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit