Cadmium, plasma standard solution, Cd 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 13813AD

Quantity 50mL

 View more versions of this product

Catalog No. AA13813AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7697-37-2,7440-43-9,7732-18-5
    Physical Form Liquid
    Molecular Formula Cd in 5% HNO3
    MDL Number MFCD00010914
    Chemical Name or Material Cadmium

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 50mL
    DOT Information Hazard Class: 8, Packing Group: III
    Health Hazard 2 When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    Appearance Clear liquid
    Molecular Formula Cd in 5% HNO3
    MDL Number MFCD00010914
    CAS 7697-37-2,7440-43-9,7732-18-5
    Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Cadmium: 0.1%; Water: 94.9%
    Health Hazard 1 H350-H314-H333: May cause cancer. Causes severe skin burns and eye damage. May be harmful if inhaled.
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Cadmium

    When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H350-H314-H333: May cause cancer. Causes severe skin burns and eye damage. May be harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit