Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cadmium rod, 10mm (0.39 in.) dia., Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 39727DJ

Quantity 50mm

 View more versions of this product

Catalog No. AA39727DJ

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-43-9
MFCD00010914
Cadmium rod, 10mm (0.39 in.) dia.
Puratronic™

Specifications

≈0.68g/mm
7440-43-9
MFCD00010914
99.9999%, (metals basis)
50mm
Rod
Cadmium rod, 10mm (0.39 in.) dia.
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit