Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium shot, 5cm (2 in.) dia., 99.9% (metals basis)
Click to view available options
Quantity:
1 ea.
Specifications
Quantity | 1 ea. |
Weight | ≈600g/shot |
Physical Form | ≈600g/shot |
Percent Purity | 99.9% |
Chemical Name or Material | Cadmium shot, 5cm (2 in.) dia. |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
H330-H341-H350-H361fd-H372-H500
EINECSNumber : 231-152-8
TSCA : Yes
Recommended Storage : Ambient temperatures