Alfa Aesar™ Cadmium shot, tear drop, -5+20 mesh, 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 1012230

Quantity 250g

 View more versions of this product

Catalog No. AA1012230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-43-9
    MDL Number MFCD00010914
    Chemical Name or Material Cadmium shot

    Specifications

    Physical Form Teardrop Shot
    Health Hazard 2 GHS H Statement
    H330-H350-H372-H341-H361
    Fatal if inhaled.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Suspected of damaging fertility or the unborn child.
    Health Hazard 3 GHS P Statement
    P260-P284-P304+P340-P320-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear respiratory protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00010914
    TSCA TSCA
    CAS 7440-43-9
    Mesh Size −5+20 Mesh
    Health Hazard 1 Danger
    Quantity 250g
    Chemical Name or Material Cadmium shot
    Assay 99.95%, (metals basis)

    GHS H Statement
    H330-H350-H372-H341-H361
    Fatal if inhaled.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P260-P284-P304+P340-P320-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear respiratory protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit