Learn More
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO3, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 195871000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Cadmium standard solution | |
1 mg/ml Cd in 0.5N HNO3 | |
10022-68-1, 7697-37-2, 7732-18-5 | |
CdH2N2O6 | |
15, 1627 | |
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O | |
238.44 | |
CHEBI:86156 | |
AAS | |
1.012 | |
Solution |
997 to 1003 μg/mL of Cd(2+) in 0.5N HNO3 (20°C) | |
Plastic bottle | |
100 mL | |
MFCD00149626 | |
IZIPRBSCGMISKX-UHFFFAOYSA-N | |
cadmium(2+) bis(nitric acid) | |
56924536 | |
308.48 | |
1.0120g/mL | |
Soluble in water |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May be corrosive to metals.
Harmful to aquatic life with long lasting effects.
May cause cancer.
GHS P Statement:
Obtain special instructions before use.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only