Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7790-84-3
CdH2O5S
226.49
VRFRVKRTIMIRNG-UHFFFAOYSA-L
cadmium(2+) hydrate sulfate
O.[Cd++].[O-]S([O-])(=O)=O
7790-84-3
CdH2O5S
226.49
VRFRVKRTIMIRNG-UHFFFAOYSA-L
cadmium(2+) hydrate sulfate
O.[Cd++].[O-]S([O-])(=O)=O

C1050, 7790-84-3, CdSO48/3H2O

Spectrum™ Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS , is used widely for the electroplating of cadmium in electronic circuits. It is also a precursor to cadmium-based pigment such as cadmium sulfide. It is also used for electrolyte in a Weston standard cell as well as a pigment in fluorescent screens. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
CAS Max % 100%
UN Number UN2570
Percent Purity 98-102%
Calcium (Ca) 0.005%
Chloride 0.001%
Copper (Cu) 0.002%
Insoluble Matter 0.005%
Lead (Pb) 0.003%
Packaging Poly Pail
Potassium (K) 0.01%
Sodium (Na) 0.02%
Zinc (Zn) 0.05%
Quantity 12 kg
Show More Show Less