Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium sulfate, ACS reagent, anhydrous, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
10124-36-4
CdO4S
208.46
QCUOBSQYDGUHHT-UHFFFAOYSA-L
cadmium sulfate,cadmium sulphate,cadmium monosulfate,sulfuric acid, cadmium salt 1:1,cadmium sulfuricum,cadmium sulfate 1:1,cadmium sulfate, tetrahydrate,cadmium 2+ sulfate,caswell no. 136c,unii-947unf3z6o
24962
CHEBI:50292
cadmium(2+);sulfate
[O-]S(=O)(=O)[O-].[Cd+2]
10124-36-4
CdO4S
208.46
QCUOBSQYDGUHHT-UHFFFAOYSA-L
cadmium sulfate,cadmium sulphate,cadmium monosulfate,sulfuric acid, cadmium salt 1:1,cadmium sulfuricum,cadmium sulfate 1:1,cadmium sulfate, tetrahydrate,cadmium 2+ sulfate,caswell no. 136c,unii-947unf3z6o
24962
CHEBI:50292
cadmium(2+);sulfate
[O-]S(=O)(=O)[O-].[Cd+2]
Linear Formula CdSO4
Merck Index 15,1632
Solubility Information Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol,acids,acetone and ammonia
Formula Weight 208.46
Grade ACS Reagent
Chloride 0.001% max
Insoluble Matter 0.005% max
Packaging Glass bottle
Solubility Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol, acids, acetone and ammonia
Melting Point 1000.0°C
Quantity 2.5kg
Chemical Name or Material Cadmium sulfate
Show More Show Less

GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
May damage fertility

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath

GHS Signal Word: Danger

EINECSNumber : 233-331-6