Alfa Aesar™ Cadmium sulfate hydrate, 98%

Manufacturer: Alfa Aesar™ A101680E

Quantity 2500g

 View more versions of this product

Catalog No. AAA101680E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15244-35-6
    Molecular Formula 3CdSO4·xH2O (x≈1)
    Formula Weight 208.46 (Anhydrous)
    MDL Number MFCD00151655
    UN Number UN2570
    Merck Index 14,1627
    Chemical Name or Material Cadmium sulfate hydrate

    Specifications

    Formula Weight 208.46 (Anhydrous)
    Health Hazard 2 GHS H Statement
    H301-H330-H340-H350-H360-H372
    Toxic if swallowed.
    Fatal if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    MDL Number MFCD00151655
    CAS 15244-35-6
    UN Number UN2570
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
    TSCA TSCA
    Molecular Formula 3CdSO4·xH2O (x≈1)
    Merck Index 14,1627
    Health Hazard 1 Danger
    RTECS Number EV2850000
    Melting Point 41.5°C (trans pt)
    Quantity 2500g
    Chemical Name or Material Cadmium sulfate hydrate
    Assay 98%

    Description

    Applications
    Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

    Solubility
    Soluble in water (1130g/L ).

    Notes
    Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.

    GHS H Statement
    H301-H330-H340-H350-H360-H372
    Toxic if swallowed.
    Fatal if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit