Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7080571
Click to view available options
Quantity:
100 g
500 g
2500 g
15244-35-6
CdO4S
208.47
MFCD00151655
QCUOBSQYDGUHHT-UHFFFAOYSA-L
cadmium(2+) sulfate
[Cd++].[O-]S([O-])(=O)=O
15244-35-6
CdO4S
208.47
MFCD00151655
QCUOBSQYDGUHHT-UHFFFAOYSA-L
cadmium(2+) sulfate
[Cd++].[O-]S([O-])(=O)=O

Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water (1130g/L ).

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 41.5°C (trans pt)
Quantity 100 g
UN Number UN2570
Merck Index 14,1627
Solubility Information Soluble in water (1130g/L ).
Formula Weight 208.46 (Anhydrous)
Percent Purity 98%
Odor Odorless
Refractive Index 1.565
Density 3.08
Chemical Name or Material Cadmium sulfate hydrate
Show More Show Less
Hazard Category H301-H330-H340-H350-H360-H372
Hazard Statement GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 233-331-6
RTECSNumber EV2850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit