Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.
Solubility
Soluble in water (1130g/L ).
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.
Solubility
Soluble in water (1130g/L ).
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Specifications
Melting Point | 41.5°C (trans pt) |
Quantity | 100 g |
UN Number | UN2570 |
Merck Index | 14,1627 |
Solubility Information | Soluble in water (1130g/L ). |
Formula Weight | 208.46 (Anhydrous) |
Percent Purity | 98% |
Odor | Odorless |
Refractive Index | 1.565 |
Density | 3.08 |
Show More |
Safety and Handling
Hazard Category | H301-H330-H340-H350-H360-H372 |
Hazard Statement | GHS H Statement H301-H330-H340-H350-H360-H372 Toxic if swallowed. Fatal if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS |
EINECSNumber | 233-331-6 |
RTECSNumber | EV2850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction