Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Sulfate, Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle
7790-84-3
CdSO4.8/3 H2O
7790-84-3
CdSO4.8/3 H2O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 256.52
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.08kg
Quantity 500g
Chemical Name or Material Cadmium Sulfate, Hydrate