Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium sulfate octahydrate, ACS, 98.0-102.0%, Alfa Aesar™

Used in Standard Weston Cell, as electrolytes in electroplating, in phosphors, in manufacture of standard cadmium elements, and as a catalyst in Marsh test for AS

Manufacturer:  Alfa Aesar 1234030

Quantity 250g

 View more versions of this product

Catalog No. AA1234030


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7790-84-3
769.50 (208.46 Anhydrous)
UN2570
Cadmium sulfate octahydrate
3CdSO4·8H2O
MFCD00151655
14,1627
ACS Grade

Specifications

7790-84-3
3CdSO4·8H2O
MFCD00151655
14,1627
3.08
1.565
>40°C (in -H2O)
Cadmium sulfate octahydrate
ACS Grade
Crystalline
769.50 (208.46 Anhydrous)
UN2570
Soluble in water. Almost insoluble in alcohol and ethyl acetate
Odorless
White
250g
98.0 to 102.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H330-H340-H350-H360-H372;
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

RTECSNumber : EV2850000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit