Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium Sulfide, Purified, Spectrum™

Click to view available options
Quantity:
125 g
500 g
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
1306-23-6
1306-23-6
Grade Purified
CAS Max % 1
Packaging Poly Pail
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.