Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cadmium wire, 0.64mm (0.025 in.) dia., Puratronic™, 99.999% (metals basis)

Click to view available options
Quantity:
15m
3m
7440-43-9
Cd
112.414
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds
23973
CHEBI:22977
cadmium
[Cd]
7440-43-9
Cd
112.414
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds
23973
CHEBI:22977
cadmium
[Cd]
Quantity 3m
Weight ≈2.8g/m
Physical Form Wire, ≈2.8g/m
Percent Purity 99.999%
Grade Puratronic™
Chemical Name or Material Cadmium wire, 0.64mm (0.025 in.) dia.
Assay (metals basis)

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c

H330-H341-H350-H361fd-H372-H500

EINECSNumber : 231-152-8

TSCA : Yes

Recommended Storage : Ambient temperatures