Learn More
CAF1 p150 Monoclonal Antibody (SS1 1-13), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA190578
Description
MA1-90578 detects CAF1B from human samples. MA1-90578 has been successfully used in immunoprecipitation, Western blot applications. The MA1-90578 immunogen is: Recombit human CAF-1p150 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B; MIM 601245), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro (Kaufman et al., 1995 [PubMed 7600578]).
Specifications
CAF1 p150 | |
Monoclonal | |
Unconjugated | |
Q13111 | |
CHAF1A | |
Recombit human CAF-1p150 | |
RUO | |
10036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
SS1 1-13 | |
CHAF1A | |
CAF-1, CAF1, CAF1B, CAF1P150, MGC71229, P150 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.