Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals

Catalog Number ac108160100
Click to view available options
Quantity:
10 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
58-08-2
C8H10N4O2
194.19
MFCD00005758
RYYVLZVUVIJVGH-UHFFFAOYSA-N
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
2519
CHEBI:27732
1,3,7-trimethylpurine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
58-08-2
C8H10N4O2
194.19
MFCD00005758
RYYVLZVUVIJVGH-UHFFFAOYSA-N
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
2519
CHEBI:27732
1,3,7-trimethylpurine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C

The crystalline powder form of the stimulant caffeine, CAS # 58-08-2, purified for analytical chemistry applications such as HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
  • It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
  • Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
TRUSTED_SUSTAINABILITY
Acidity 0.2mL of 0.01M NaOH max.
Melting Point 234.0°C to 239.0°C
Color White
Density 1.2300g/mL
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. (total impurities) (HPLC)
Packaging Glass Bottle
Quantity 10 g
Beilstein 26,461
Merck Index 15,1639
Specific Gravity 1.23
Solubility Information Solubility in water: 20g/l (20°C). Other solubilities: 1g/66 mL alcohol,1g/50 mL acetone,freely soluble in pyrrole,soluble in ethyl acetate
Sublimation Point 178 °C
Residual Solvents meets the requirements
Formula Weight 194.19
Percent Purity 98.5%
Related Substances 0.5% max. (TLC)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Caffeine, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-362-1
RTECSNumber EV6475000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit