Learn More
Caffeine, Anhydrous, FCC, 98.5-101.%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
C1056, 58-08-2, C8H10N4O2
Spectrum™ Caffeine, Anhydrous, FCC is used as a flavoring ingredient in a variety of beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle