Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeine, Anhydrous, FCC, 98.5-101.%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
58-08-2
C8H10N4O2
194.19
RYYVLZVUVIJVGH-UHFFFAOYSA-N
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
58-08-2
C8H10N4O2
194.19
RYYVLZVUVIJVGH-UHFFFAOYSA-N
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C

C1056, 58-08-2, C8H10N4O2

Spectrum™ Caffeine, Anhydrous, FCC is used as a flavoring ingredient in a variety of beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 2.5 kg
Ignition Residue 0.1%
Water 0.5%
Melting Range 235-237.5°C
Packaging Poly Pail
CAS Max % 100%
UN Number UN1544
Percent Purity 98.5-101.%