Learn More
Caffeine, Anhydrous, USP, 98.5-101%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
CA105, 58-08-2, C8H10N4O2
Spectrum™ Caffeine, Anhydrous, USP is a central nervous system (CNS) stimulant of the methylxanthine class used in some migraine medications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.001
Packaging Poly Bottle