Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caffeine, Anhydrous, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
58-08-2
C8H10N4O2
194.19
RYYVLZVUVIJVGH-UHFFFAOYSA-N
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
58-08-2
C8H10N4O2
194.19
RYYVLZVUVIJVGH-UHFFFAOYSA-N
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C

CA105, 58-08-2, C8H10N4O2

Spectrum™ Caffeine, Anhydrous, USP is a central nervous system (CNS) stimulant of the methylxanthine class used in some migraine medications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN1544
Ignition Residue 0.1%
Percent Purity 98.5-101%
Packaging Poly Pail
Quantity 2.5 kg