Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calamine, Powder, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
8011-96-9
8011-96-9

CA115, 8011-96-9

Spectrum™ Calamine, Powder, USP is used in topical medications to relieve pain and itching. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Quantity 500 g
CAS Max % 1
Percent Purity 98 to 100.5%