Calapro Antistatic Latex Finger Cots
Antistatic, super-thin.
Manufacturer: CALAPRO INC FW04S
 View all options for this product
Catalog No. 11-392-16A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Made of 100% pure, super-thin latex (5mil thick) for comfort and dexterity. Fit without bulkiness at the tip to reduce finger fatigue.
    • For ISO Class 5 (formerly Class 100) applications
    • Average surface resistivity of 1012 ohms/cm2
    • Use when handling Class II Category B (≥1000V) static-sensitive devices
    • Measure 2 3/4 in. long (7cm)
  • Description & Specifications
Related Products
Product Content Feedback