Calapro Antistatic Latex Finger Cots

Antistatic, super-thin.

Manufacturer: CALAPRO INC FW04L

 View more versions of this product

Catalog No. 11-392-16C

  Check Availability
Add to Cart

Alert

Warning: The products featured on this page contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


    Specifications

    Certifications/Compliance ISO Class 5
    Color Pink
    For Use With (Application) For static-free areas
    For Use With (Equipment) Used when handling Class II category B (=1000V) static-sensitive devices
    Length (English) 2.75 in.
    Length (Metric) 7cm
    Material Natural Rubber latex
    Powder-Free Yes
    Size Large
    Thickness (English) 5mil
    Product Type Antistatic Finger Cot

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit