Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calapro Powder-Free Latex Finger Cots

Made of 100% pure, super-thin latex (5mil thick) for comfort and dexterity

Manufacturer:  CALAPRO INC FY01L

 View more versions of this product

Catalog No. 11-392-9C



  Check Availability
Add to cart

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Specifications Description & Specifications

Specifications

ISO class 5
0.78 in.
For comfort and dexterity
7cm
Yes
4.5
Finger Cot
Ember
2cm
2.75 in.
Natural Latex
Large
5mil
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit