Learn More
Calbindin D-28K clone 1A11 Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 1A11, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB177825UL
 View more versions of this product
Catalog No. ZRB177825MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Calbindin D-28K clone 1A11 Monoclonal antibody specifically detects Calbindin D-28K clone 1A11 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Calbindin D-28K clone 1A11
1A11
Unconjugated
Calbindin;D28;D-28K;PCD-29;Spot 35 protein;Vitamin D-dependent calcium-binding protein avian-type
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
Human, Mouse, Rat
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
Calb1
GST/His-tagged recombinant fragment corresponding to 107 amino acids from the C-terminal half of murine Calbindin.
∼28 kDa observed; 29.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal