Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calbindin D-28K clone 1A11 Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 1A11, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Calbindin D-28K clone 1A11
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB177825MI
View Documents
MilliporeSigma
ZRB177825UL
25 μL N/A N/A N/A
ZRB17784XMI
View Documents
MilliporeSigma
ZRB17784X25UL
4 x 25 μL N/A N/A N/A
Description

Description

Calbindin D-28K clone 1A11 Monoclonal antibody specifically detects Calbindin D-28K clone 1A11 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

Calbindin D-28K clone 1A11
Unconjugated
Rabbit
Calbindin;D28;D-28K;PCD-29;Spot 35 protein;Vitamin D-dependent calcium-binding protein avian-type
GST/His-tagged recombinant fragment corresponding to 107 amino acids from the C-terminal half of murine Calbindin.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
Calb1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit