Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calbindin D-28K Mouse anti-Human, Clone: CALB1/2782, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30720810UG

 View more versions of this product

Catalog No. NB121889


Add to cart

Description

Description

Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Calbindin D-28K
CALB1/2782
Unconjugated
10 mM PBS with 0.05% BSA
CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
ELISA, Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG2b Kappa
100 μg
Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary
793
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit