Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calbindin D-28K Mouse anti-Human, Clone: CALB1/3333, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Calbindin D-28K
Clone CALB1/3333
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications ELISA, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122201
View Documents
Novus Biologicals
NBP30736410UG
100 μg
Each for $352.89
Add to cart
 
NB122202
View Documents
Novus Biologicals
NBP30736420UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Calbindin D-28K
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
CALB1/3333
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
10 mM PBS with 0.05% BSA
793
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit