Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ 1-Deoxymannojirimycin, Hydrochloride
A specific α-mannosidase I inhibitor that blocks conversion of high mannose to complex oligosaccharides.
Supplier: MilliporeSigma™ 2605755MG
Description
Packaged Under Inert GasSpecifications
Crystalline Solid | |
5 mg | |
98% by TLC | |
84444-90-6 | |
199.6g/mol |
Hygroscopic | |
White | |
α-mannosidase-I | |
C6H13NO4·HCl | |
EtOH or H2O |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title