Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ 1-Deoxynojirimycin, Hydrochloride
A specific glucosidase inhibitor including trimming glucosidases I and II, enzymes that sequentially remove the three glucose residues from precursor Glc3
Supplier: MilliporeSigma™ 26068410MG
Description
A specific glucosidase inhibitor including trimming glucosidases I and II, enzymes that sequentially remove the three glucose residues from precursor Glc3Man9-GlcNAc2 in N-linked glycan biosynthesis.Specifications
Hygroscopic | |
Crystalline Solid | |
Glucosidase 1, glucosidase 2 | |
C6H13NO4·HCl | |
H2O |
10 mg | |
98% by TLC | |
73285-50-4 | |
199.6g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title