Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 4-{[3',4'-(Methylenedioxy)benzyl]amino}-6-methoxyquinazoline

Catalog No. 4752505MG
Click to view available options
Quantity:
5 mg

Potent and specific inhibitor of cGMP-specific phosphodiesterase (PDE V; IC50 = 230

Potent and specific inhibitor of cGMP-specific phosphodiesterase (PDE V; IC50 = 230nM). Elevates the intracellular cGMP level without causing any change in the cAMP level in isolated porcine coronary arteries. Has no effect on other PDE isozymes.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins CGMP-PDEV
Molecular Formula C17H15N3O3
Formula Weight 309.3g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.