Learn More
MilliporeSigma™ Calbiochem™ 7S NGF, Mouse Submaxillary Glands
Helps in regulation of cell growth, replication, and differentiation
Manufacturer:  MilliporeSigma™ 480354100UG
Catalog No. 48-035-4100UG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Promotes neuron survival and neurite outgrowth in newborn rat brain
  • Composed of three subunits: (α:β:γ): 2:1:2
  • α-subunit is inactive serine proteinase
  • γ-subunit is active serine proteinase capable of processing precursor form of β-NGF
Specifications
Specifications
Lyophilized
100μg
7S Nerve Growth Factor
Lyophilized from sterile water.
2° to 8°C
7S NGF, Mouse Submaxillary Glands