Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ A77 1726

Blocks TNF-dependent NF-κB activation gene expression, inhibits activity of dihydroorotate dehydrogenase.

Manufacturer:  MilliporeSigma™ 1001285MG

Catalog No. 10-012-85MG


Add to cart

Description

Description

  • Cell-permeable, reversible, substrate competitive inhibitor that inhibits IL-2-induced tyrosine phosphorylation of JAK1 JAK3
  • Reported to reduce IFN-γ-induced expression of iNOS in L929 fibroblasts
  • Inhibits IL-1β-induced activation of COX-2 PGE2 synthesis (IC50 = 130nM) in A549 cells
  • Assay: ≥97% by HPLC
  • CAS: 108605-62-5
  • Molecular Formula: C12H9F3N2O2
  • Color: White
Specifications

Specifications

2°C to 8°C
5 mg
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit