Learn More
MilliporeSigma™ Calbiochem™ Acetyl-11-keto-β-Boswellic Acid, Boswellia serrata
A cell-permeable pentacyclic triterpene of the ursane type that displays anti-tumor and anti-inflammatory properties.
Manufacturer:  MilliporeSigma™ 1101235MG
Catalog No. 11-012-35MG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
98% by HPLC
Protect from Light
5-lipoxygenase
C32H48O5
DMSO
White solid
5mg
67416-61-9
512.7g/mol