EMD Millipore™ Calbiochem™ Acetylcoenzyme A, Trilithium Salt Available on GSA/VA Contract for Federal Government customers only.
Carrier of acyl groups in enzymatic reactions in fatty acid synthesis and oxidation and in biological acetylations
Manufacturer: EMD Millipore™ 106310MG
 View more versions of this product
Catalog No. 10-631-0MG
  Check Availability
Add to Cart
    Specifications
    Additive Acetylcoenzyme A, Trilithium Salt
    Quantity 10mg
    Storage Requirements -20°C
  • Description & Specifications
Related Products
Product Content Feedback