Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Acridine Orange

Cell-permeable, cationic fluorescent dye that Interacts with DNA RNA by intercalation or electrostatic attractions

Manufacturer:  MilliporeSigma™ 1130001GM

Catalog No. 11-300-01GM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Associates with RNA, excitation maximum is 460nm, emission maximum is 650nm (red)
  • Used to measure single, double-stranded DNA RNA in diagnosis, classification, prognosis of many neoplasms
  • Versatile fluorescent stain
  • Used in histochemistry cytochemistry to underst about in situ content, molecular structure, conformation of many nucleic acid-containing cell constituents
  • Assay: ≥98% by HPLC
  • CAS: 65-61-2
  • Molecular Formula: C17H20ClN3
Specifications

Specifications

Burnt-Orange
DNA and RNA
−15° to 30°C
Acridine Orange
1g
Safety and Handling

Safety and Handling

ShelfLife : Aqueous stock solutions are Stable for up to 1 month at 4°C

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit