Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Calphostin C, Cladosporium cladosporioides
A cell permeable, highly specific inhibitor of protein kinase C (IC50 = 50nM) that interacts with the protein's regulatory domain by competing at the binding site of diacylglycerol and phorbol esters.
Supplier: MilliporeSigma™ 208725100UG
Description
A cell permeable, highly specific inhibitor of protein kinase C (IC50 = 50nM) that interacts with the protein’s regulatory domain by competing at the binding site of diacylglycerol and phorbol esters. Does not compete with Ca2+ or phospholipids. At higher concentrations inhibits myosin light chain kinase (IC50 > 5μM), protein kinase A (IC50 > 50μM), protein kinase G (IC50 > 25μM), and p60v- src (IC50 > 50μM). Induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells. Requires brief exposure to light for activation.Specifications
Lyophilized Solid | |
100 μg | |
95% by HPLC | |
121263-19-2 | |
790.8g/mol |
Protect from light | |
Light Brown | |
PKC | |
C44H38O14 | |
DMSO or EtOH |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title